Learn More
Medchemexpress LLC Boc-Pen(Mob)-OH | 120944-75-4 | MFCD00076975 | 99.9% | 369.48 g/mol | C18H27NO5S | 5 G
SDP
Catalog No. 5000283448
Encompass_Preferred
Boc-Pen(Mob)-OH is a Boc-protected penicillamine derivative used as an amino acid building block in peptide synthesis and other research applications. It is supplied as a white to off-white solid and is available in small laboratory pack sizes for standard research workflows.
  • Used as an amino acid building block for peptide synthesis and modification
  • Molecular formula C18H27NO5S and molecular weight 369.48 g/mol
  • High purity suitable for research applications
  • Available in multiple pack sizes for lab-scale use
  • Stored refrigerated; solutions benefit from low-temperature storage under inert gas
Catalog No. 50-002-83448 Supplier Medchemexpress LLC Supplier No. HYW1418155G
Boc-Pen(Mob)-OH is a Boc-protected penicillamine derivative used as an amino acid building block in peptide synthesis and other research applications. It is supplied as a white to off-white solid and is available in small laboratory pack sizes for standard research workflows.
  • Used as an amino acid building block for peptide synthesis and modification
  • Molecular formula C18H27NO5S and molecular weight 369.48 g/mol
  • High purity suitable for research applications
  • Available in multiple pack sizes for lab-scale use
  • Stored refrigerated; solutions benefit from low-temperature storage under inert gas