Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Phe(4-Cl)-OH | 68090-88-0 | 299.75 | 25 G
SDP

Catalog No. 5000448234
Encompass_Preferred

Boc-Phe(4-Cl)-OH is an amino acid derivative, specifically a phenylalanine derivative. Amino acid derivatives like this product are recognized for their potential benefits as ergogenic dietary substances. They can influence the secretion of anabolic hormones, contribute to fuel supply during exercise, improve mental performance during stress-related tasks, and help prevent exercise-induced muscle damage.

  • Phenylalanine derivative.
  • Can influence the secretion of anabolic hormones.
  • Contributes to fuel supply during exercise.
  • May improve mental performance during stress-related tasks.
  • Helps prevent exercise-induced muscle damage.
  • Appearance: Solid, white to off-white.
  • Purity: 99.75%.
  • Molecular weight: 299.75.
  • Formula: C14H18ClNO4.

Catalog No. 50-004-48234 Supplier Medchemexpress LLC Supplier No. 25GHYW008486
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Phe(4-Cl)-OH is an amino acid derivative, specifically a phenylalanine derivative. Amino acid derivatives like this product are recognized for their potential benefits as ergogenic dietary substances. They can influence the secretion of anabolic hormones, contribute to fuel supply during exercise, improve mental performance during stress-related tasks, and help prevent exercise-induced muscle damage.

  • Phenylalanine derivative.
  • Can influence the secretion of anabolic hormones.
  • Contributes to fuel supply during exercise.
  • May improve mental performance during stress-related tasks.
  • Helps prevent exercise-induced muscle damage.
  • Appearance: Solid, white to off-white.
  • Purity: 99.75%.
  • Molecular weight: 299.75.
  • Formula: C14H18ClNO4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.