Learn More
Medchemexpress LLC Boc-Phe-Leu-Phe-Leu-Phe | 73572-58-4 | 98.0% | 785.97 | 10 MG
SDP
Catalog No. 5000397303
Encompass_Preferred
Boc-Phe-Leu-Phe-Leu-Phe (Boc-FLFLF) is an N-formyl peptide receptors (FPR) inhibitor. It abolishes the FMLP-induced release of peptide leukotrienes. This compound inhibits the sprouting of human umbilical vein endothelial cell (HUVEC) spheroids mediated by proliferative diabetic retinopathy (PDR) vitreous and vascular endothelial growth factor (VEGF) respectively in a three-dimensional fibrin gel. Boc-Phe-Leu-Phe-Leu-Phe also inhibits the anti-inflammatory and antifibrotic effects of Ac2-26 and can be used for the study of immunology.
  • Inhibits n-formyl peptide receptors (FPR)
  • Abolishes FMLP-induced release of peptide leukotrienes
  • Inhibits sprouting of human umbilical vein endothelial cell (HUVEC) spheroids
  • Inhibits anti-inflammatory and antifibrotic effects of Ac2-26
  • Applicable for immunology studies
Catalog No. 50-003-97303 Supplier Medchemexpress LLC Supplier No. HYP331810MG
Boc-Phe-Leu-Phe-Leu-Phe (Boc-FLFLF) is an N-formyl peptide receptors (FPR) inhibitor. It abolishes the FMLP-induced release of peptide leukotrienes. This compound inhibits the sprouting of human umbilical vein endothelial cell (HUVEC) spheroids mediated by proliferative diabetic retinopathy (PDR) vitreous and vascular endothelial growth factor (VEGF) respectively in a three-dimensional fibrin gel. Boc-Phe-Leu-Phe-Leu-Phe also inhibits the anti-inflammatory and antifibrotic effects of Ac2-26 and can be used for the study of immunology.
  • Inhibits n-formyl peptide receptors (FPR)
  • Abolishes FMLP-induced release of peptide leukotrienes
  • Inhibits sprouting of human umbilical vein endothelial cell (HUVEC) spheroids
  • Inhibits anti-inflammatory and antifibrotic effects of Ac2-26
  • Applicable for immunology studies