Learn More
Bachem Boc-Phe-Ser-Arg-AMC Acetate
SDP
Catalog No. NC1807687
Encompass_Preferred
Boc-Phe-Ser-Arg-AMC Acetate, 73554-90-2, C33H43N7O8, Mr 665.75, 5mg. The fluorogenic substrate Boc-FSR-AMC is preferentially hydrolyzed by coagulation factor XIa. Boc-FSR-AMC is also highly susceptible to the proteases in yeast mitochondrial matrix as well as to tryptase from rat mast cells. In addition, the substrate is cleaved by acrosin from the ascidian Halocynthia roretzi. Stock solutions of Boc-FSR-AMC are best prepared in DMSO.
Catalog No. NC1807687 Supplier Bachem Supplier No. 4012340.0005
Add to Cart
Add to Cart
Boc-Phe-Ser-Arg-AMC Acetate, 73554-90-2, C33H43N7O8, Mr 665.75, 5mg. The fluorogenic substrate Boc-FSR-AMC is preferentially hydrolyzed by coagulation factor XIa. Boc-FSR-AMC is also highly susceptible to the proteases in yeast mitochondrial matrix as well as to tryptase from rat mast cells. In addition, the substrate is cleaved by acrosin from the ascidian Halocynthia roretzi. Stock solutions of Boc-FSR-AMC are best prepared in DMSO.