Learn More
Medchemexpress LLC (±)-Boc-α-phosphonoglycine trimethyl ester | 89524-98-1 | 297.24 | 25 G
SDP
Catalog No. 5000448178
Encompass_Preferred
(±)-Boc-α-phosphonoglycine trimethyl ester serves as a glycine equivalent and donor in organic synthesis. It is notable for its strong α-proton acidity and water-soluble reaction byproducts, which simplify purification.
  • Serves as a glycine equivalent
  • Functions as a glycine donor in organic synthesis
  • Possesses strong α-proton acidity
  • Reaction byproducts are highly water-soluble
  • Simplifies purification processes
  • Enables efficient synthesis of target 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieves synthesis through four mild reactions
Catalog No. 50-004-48178 Supplier Medchemexpress LLC Supplier No. 25GHYW008206
(±)-Boc-α-phosphonoglycine trimethyl ester serves as a glycine equivalent and donor in organic synthesis. It is notable for its strong α-proton acidity and water-soluble reaction byproducts, which simplify purification.
  • Serves as a glycine equivalent
  • Functions as a glycine donor in organic synthesis
  • Possesses strong α-proton acidity
  • Reaction byproducts are highly water-soluble
  • Simplifies purification processes
  • Enables efficient synthesis of target 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieves synthesis through four mild reactions