Learn More
Medchemexpress LLC (±)-Boc-α-phosphonoglycine trimethyl ester | 89524-98-1 | 297.24 | 5 G
SDP
Catalog No. 5000448528
Encompass_Preferred
(±)-Boc-α-phosphonoglycine trimethyl ester is a chemical compound that functions as a glycine equivalent and glycine donor in organic synthesis. It is characterized by strong α-proton acidity, and its reaction byproducts are highly water-soluble, facilitating easy purification. This compound is effectively used to obtain 5,6- and 5,8-dimethoxy-substituted ester products through a sequence of four mild reactions.
  • Serves as a glycine equivalent and glycine donor
  • Exhibits strong alpha-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Efficiently obtains 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieves results via four mild reactions
Catalog No. 50-004-48528 Supplier Medchemexpress LLC Supplier No. HYW0082065G
(±)-Boc-α-phosphonoglycine trimethyl ester is a chemical compound that functions as a glycine equivalent and glycine donor in organic synthesis. It is characterized by strong α-proton acidity, and its reaction byproducts are highly water-soluble, facilitating easy purification. This compound is effectively used to obtain 5,6- and 5,8-dimethoxy-substituted ester products through a sequence of four mild reactions.
  • Serves as a glycine equivalent and glycine donor
  • Exhibits strong alpha-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Efficiently obtains 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieves results via four mild reactions