Learn More
Medchemexpress LLC (±)-Boc-α-phosphonoglycine trimethyl ester | 89524-98-1 | 297.24 | 50 G
SDP
Catalog No. 5000448790
Encompass_Preferred
(±)-Boc-α-phosphonoglycine trimethyl ester functions as a glycine equivalent and donor in organic synthesis. Its reaction byproducts are water-soluble and easily purified. It facilitates the efficient synthesis of 5,6- and 5,8-dimethoxy-substituted ester products via four mild reactions.
  • Serves as a glycine equivalent and glycine donor
  • Possesses strong α-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Used to efficiently obtain 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieved through four mild reactions
Catalog No. 50-004-48790 Supplier Medchemexpress LLC Supplier No. HYW00820650G
(±)-Boc-α-phosphonoglycine trimethyl ester functions as a glycine equivalent and donor in organic synthesis. Its reaction byproducts are water-soluble and easily purified. It facilitates the efficient synthesis of 5,6- and 5,8-dimethoxy-substituted ester products via four mild reactions.
  • Serves as a glycine equivalent and glycine donor
  • Possesses strong α-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Used to efficiently obtain 5,6- and 5,8-dimethoxy-substituted ester products
  • Achieved through four mild reactions