Learn More
Medchemexpress LLC (±)-Boc-α-phosphonoglycine trimethyl ester | 89524-98-1 | MFCD02178893 | 297.24 | 10 G
SDP
Catalog No. 5000449394
Encompass_Preferred
(±)-Boc-α-phosphonoglycine trimethyl ester (MedChemExpress HY-W008206) is a chemical compound used as a glycine equivalent and glycine donor in organic synthesis. It has strong α-proton acidity and its reaction byproducts are water-soluble and easy to purify.
  • Functions as a glycine equivalent and glycine donor
  • Strong α-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Efficiently yields target 5,6- and 5,8-dimethoxy-substituted ester products
  • Used in four mild reactions
  • For research use only
Catalog No. 50-004-49394 Supplier Medchemexpress LLC Supplier No. HYW00820610G
(±)-Boc-α-phosphonoglycine trimethyl ester (MedChemExpress HY-W008206) is a chemical compound used as a glycine equivalent and glycine donor in organic synthesis. It has strong α-proton acidity and its reaction byproducts are water-soluble and easy to purify.
  • Functions as a glycine equivalent and glycine donor
  • Strong α-proton acidity
  • Reaction byproducts are water-soluble and easy to purify
  • Efficiently yields target 5,6- and 5,8-dimethoxy-substituted ester products
  • Used in four mild reactions
  • For research use only