Learn More
Medchemexpress LLC Boc-Pip-butyn | 1346153-07-8 | 95.79% | 237.34 | 25 MG
SDP
Catalog No. 5000363290
Encompass_Preferred
Boc-Pip-butyn is an alkyl/ether-based PROTAC linker that functions as a click chemistry reagent. It incorporates an alkyne group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • It is an alkyl/ether-based PROTAC linker.
  • It serves as a click chemistry reagent.
  • It contains an alkyne group.
  • It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • It is intended for research use only.
Catalog No. 50-003-63290 Supplier Medchemexpress LLC Supplier No. 25MGHY147052
Boc-Pip-butyn is an alkyl/ether-based PROTAC linker that functions as a click chemistry reagent. It incorporates an alkyne group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing azide groups.
  • It is an alkyl/ether-based PROTAC linker.
  • It serves as a click chemistry reagent.
  • It contains an alkyne group.
  • It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • It is intended for research use only.