Learn More
Medchemexpress LLC Boc-pip-butyn | 1346153-07-8 | 95.8% | C14H23NO2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14705210MG
Encompass_Preferred
Boc-Pip-butyn is an alkyl/ether-based PROTAC linker and click-chemistry reagent containing an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is intended for introducing alkyne functionality in PROTAC assembly and bioconjugation workflows.
  • Alkyne handle compatible with CuAAC reactions.
  • Designed for PROTAC linker synthesis and bioconjugation.
  • High purity (95.8%) for consistent reaction performance.
  • Available in multiple small research quantities; supplied as a colorless to light yellow oil.
Catalog No. 50-002-61831