Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-piperazine-OH | 77279-24-4 | 99.55% | 230.30 | 500 G
SDP

Catalog No. 5000401352
Encompass_Preferred

Boc-Piperazine-OH is a PROTAC Linker. It is for research use only. This compound is suitable for various laboratory applications requiring a linker.

  • Colorless to light yellow appearance
  • Solid below 39°C, liquid above 39°C
  • Recommended storage for pure form: -20°C for up to 3 years, 4°C for up to 2 years
  • Recommended storage in solvent: -80°C for up to 6 months, -20°C for up to 1 month

Catalog No. 50-004-01352 Supplier Medchemexpress LLC Supplier No. HY20797500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Piperazine-OH is a PROTAC Linker. It is for research use only. This compound is suitable for various laboratory applications requiring a linker.

  • Colorless to light yellow appearance
  • Solid below 39°C, liquid above 39°C
  • Recommended storage for pure form: -20°C for up to 3 years, 4°C for up to 2 years
  • Recommended storage in solvent: -80°C for up to 6 months, -20°C for up to 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.