Learn More
Apexbio Technology LLC Boc-Pro-OH 15761-39-4 100g
SDP
Catalog No. 5000094162
Encompass_Preferred
Boc-Pro-OH (15761-39-4) is a t-butoxycarbonyl (Boc)-protected derivative of the amino acid proline Boc-Pro-OH serves as an amino-protecting reagent in peptide synthesis facilitating the construction and sequential elongation of peptide chains The Boc group can be selectively removed under specific reaction conditions enabling efficient amino acid coupling reactions Based on these properties Boc-Pro-OH holds research potential in the synthesis of peptide-based molecules for studying peptide structure-function relationships and assembling substrates or probes to investigate protein conformational dynamics and interaction mechanisms
Catalog No. 50-000-94162 Supplier Apexbio Technology LLC Supplier No. A7815100000
Boc-Pro-OH (15761-39-4) is a t-butoxycarbonyl (Boc)-protected derivative of the amino acid proline Boc-Pro-OH serves as an amino-protecting reagent in peptide synthesis facilitating the construction and sequential elongation of peptide chains The Boc group can be selectively removed under specific reaction conditions enabling efficient amino acid coupling reactions Based on these properties Boc-Pro-OH holds research potential in the synthesis of peptide-based molecules for studying peptide structure-function relationships and assembling substrates or probes to investigate protein conformational dynamics and interaction mechanisms