Learn More
Medchemexpress LLC Boc-propargylamine | 92136-39-5 | 99.5% | C8H13NO2 | 10 G
SDP
Catalog No. 5000434321
Encompass_Preferred
N-Boc-Propargylamine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is a click chemistry reagent, containing an alkyne group, that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used to prepare triazolobenzylidene-thiazolopyrimidines
  • Acts as CDC25 phosphatase inhibitors
  • Used for the synthesis of beta-glucan polysaccharide analogs
  • Involved in the Pauson-Khand (PK) reaction to prepare 4,5-dissubstituted cyclopentenones
Catalog No. 50-004-34321 Supplier Medchemexpress LLC Supplier No. HY7597010G
N-Boc-Propargylamine is a biochemical reagent used as a biological material or organic compound for life science-related research. It is a click chemistry reagent, containing an alkyne group, that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used to prepare triazolobenzylidene-thiazolopyrimidines
  • Acts as CDC25 phosphatase inhibitors
  • Used for the synthesis of beta-glucan polysaccharide analogs
  • Involved in the Pauson-Khand (PK) reaction to prepare 4,5-dissubstituted cyclopentenones