Learn More
Medchemexpress LLC Boc-Propargylamine | 92136-39-5 | 99.5% | C8H13NO2 | 25 G
SDP
Catalog No. 5000358510
Encompass_Preferred
Boc-Propargylamine is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used to prepare triazolobenzylidene-thiazolopyrimidines, which act as CDC25 phosphatase inhibitors.
  • Used for the synthesis of beta-glucan polysaccharide analogs.
  • Involved in the Pauson-Kh reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-dissubstituted cyclopentenones.
Catalog No. 50-003-58510 Supplier Medchemexpress LLC Supplier No. 25GHY75970
Boc-Propargylamine is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • Used to prepare triazolobenzylidene-thiazolopyrimidines, which act as CDC25 phosphatase inhibitors.
  • Used for the synthesis of beta-glucan polysaccharide analogs.
  • Involved in the Pauson-Kh reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-dissubstituted cyclopentenones.