Learn More
Medchemexpress LLC Boc-Propargylamine | 92136-39-5 | 99.78% | C8H13NO2 | 100 G
SDP
Catalog No. 5000433840
Encompass_Preferred
N-Boc-Propargylamine is a biochemical reagent and organic compound for life science research. It serves as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Prepares triazolobenzylidene-thiazolopyrimidines, which act as CDC25 phosphatase inhibitors.
  • Synthesizes beta-glucan polysaccharide analogs.
  • Involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to create 4,5-dissubstituted cyclopentenones.
Catalog No. 50-004-33840 Supplier Medchemexpress LLC Supplier No. HY75970100G
N-Boc-Propargylamine is a biochemical reagent and organic compound for life science research. It serves as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Prepares triazolobenzylidene-thiazolopyrimidines, which act as CDC25 phosphatase inhibitors.
  • Synthesizes beta-glucan polysaccharide analogs.
  • Involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to create 4,5-dissubstituted cyclopentenones.