Learn More
Bachem Boc-Gln-Ala-Arg-AMC · HCl
SDP
Supplier:  Bachem 4017019.0025
Encompass_Preferred
Boc-Gln-Ala-Arg-AMC · HCl, 201849-55-0, C29H42N8O8 · HCl, Mr 667.16, 25mg. Boc-QAR-AMC, highly reactive substrate for trypsin. Fluorogenic substrate for prostasin and matriptase.
Catalog No. 50-194-7056