Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC SCIENCES BOC Sciences Vancomycin-[d10] Monoacetate, >98% Purity, Labelled Salt, 1 mg

Catalog No. NC3950533
Encompass

Vancomycin-[d10] monoacetate salt is a labeled acetate form of vancomycin used as a potent antimicrobial agent. It functions by interfering with bacterial cell wall synthesis through binding to D-alanine-D-alanine residues, demonstrating efficacy against Gram-positive bacteria including MRSA.

  • Labeled with d10 for analytical applications
  • High chemical purity of >98%
  • Molecular weight of 1519.38 g/mol
  • Molecular formula C68H69D10Cl2N9O26
  • Active against Gram-positive bacteria and MRSA
  • Mechanism involves binding to D-alanine-D-alanine residues

Catalog No. NC3950533 Supplier BOC SCIENCES Supplier No. BLP0075711MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vancomycin-[d10] monoacetate salt is a labeled acetate form of vancomycin used as a potent antimicrobial agent. It functions by interfering with bacterial cell wall synthesis through binding to D-alanine-D-alanine residues, demonstrating efficacy against Gram-positive bacteria including MRSA.

  • Labeled with d10 for analytical applications
  • High chemical purity of >98%
  • Molecular weight of 1519.38 g/mol
  • Molecular formula C68H69D10Cl2N9O26
  • Active against Gram-positive bacteria and MRSA
  • Mechanism involves binding to D-alanine-D-alanine residues

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.