Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boc-Thr(Fmoc-Ala)-OH

Catalog No. CDS005133 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005133 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005133 Supplier Sigma Aldrich Supplier No. CDS005133100MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pd/sio2 modified with ionic liquid catalyzed lead free stereoselective hydrogenation of 2-hexyne has been reported
  • Polymerization of 2-hexyne in toluene catalyzed by mocl5.ph4sn and wcl6.ph4sn has been reported
  • Mechanism of hydrogenation of 2-hexyne in solution on selective and unselective bulk pd has been investigated
  • Lanthanum-catalyzed dimerization of 2-hexyne has been reported

Specifications

Molecular Formula C27H32N2O8
Linear Formula C27H32N2O8
MDL Number MFCD10001359
Quantity 5 g
Molecular Weight (g/mol) 512.55
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.