Learn More
Description
- The gas-phase reactions of cl atoms with methyl difluoroacetate have been investigated at 296 +/- 2k and atmospheric pressure of synthetic air.methyl difluoroacetate has been used:
- To compose the electrolyte for lithium ion cells, also showed good inhibition of aluminum cathode current collector corrosion
- As electrolyte solvent for improvement of thermal stability in lithium-ion batteries
Specifications
Specifications
| Molecular Formula | C30H38N2O8 |
| Linear Formula | C30H38N2O8 |
| MDL Number | MFCD11974989 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 554.63 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.