Learn More
Medchemexpress LLC HY-W038702 500mg Medchemexpress, Boc-Val-Cit-OH CAS:870487-08-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW038702500MG
Encompass_Preferred
Medchemexpress, HY-W038702 500mg Boc-Val-Cit-OH CAS:870487-08-4 Boc-Val-Cit-OH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3454