Learn More
Medchemexpress LLC Boc-Val-Cit-PAB | 870487-09-5 | MFCD22381208 | 99.6% | 479.57 g/mol | C23H37N5O6 | 250 MG
SDP
Catalog No. 5000203560
Encompass_Preferred
Boc-Val-Cit-PAB is a cleavable peptide-based ADC linker that uses a valine-citrulline protease-cleavable dipeptide and a para-aminobenzyl (PAB) self-immolative spacer to release payloads after internalization. It is supplied as a powder with high purity for use in antibody-drug conjugate synthesis.
  • Cleavable valine-citrulline linker enabling enzyme-triggered payload release.
  • Para-aminobenzyl (PAB) self-immolative spacer for efficient payload liberation.
  • Boc-protected N-terminus for stability during conjugation workflows.
  • High purity suitable for research-grade ADC synthesis.
  • Available in multiple pack sizes, including 250 mg.
  • Store powder at -20°C for long-term stability, aliquot solutions to avoid freeze-thaw.
Catalog No. 50-002-03560 Supplier Medchemexpress LLC Supplier No. HY141141250MG
Boc-Val-Cit-PAB is a cleavable peptide-based ADC linker that uses a valine-citrulline protease-cleavable dipeptide and a para-aminobenzyl (PAB) self-immolative spacer to release payloads after internalization. It is supplied as a powder with high purity for use in antibody-drug conjugate synthesis.
  • Cleavable valine-citrulline linker enabling enzyme-triggered payload release.
  • Para-aminobenzyl (PAB) self-immolative spacer for efficient payload liberation.
  • Boc-protected N-terminus for stability during conjugation workflows.
  • High purity suitable for research-grade ADC synthesis.
  • Available in multiple pack sizes, including 250 mg.
  • Store powder at -20°C for long-term stability, aliquot solutions to avoid freeze-thaw.