Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-val-cit-pab-pnp | 870487-10-8 | 99.6% | 644.67 | C30H40N6O10 | 50 MG
SDP

Catalog No. 5000203263
Encompass_Preferred

Boc-Val-Cit-PAB-PNP is a protease-cleavable linker used to construct antibody-drug conjugates (ADCs). It releases payloads after intracellular proteolytic cleavage and is supplied as a solid with high purity.

  • Cleavable val-cit peptide linker releasing payload upon proteolysis
  • Para-aminobenzyl spacer with p-nitrophenyl leaving group
  • High purity suitable for research applications
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance
  • Recommended storage: -20°C under nitrogen; in solvent store at -80°C for long-term
  • Available in small research scales, e.g. 50 mg

Catalog No. 50-002-03263 Supplier Medchemexpress LLC Supplier No. HY14114250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Val-Cit-PAB-PNP is a protease-cleavable linker used to construct antibody-drug conjugates (ADCs). It releases payloads after intracellular proteolytic cleavage and is supplied as a solid with high purity.

  • Cleavable val-cit peptide linker releasing payload upon proteolysis
  • Para-aminobenzyl spacer with p-nitrophenyl leaving group
  • High purity suitable for research applications
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance
  • Recommended storage: -20°C under nitrogen; in solvent store at -80°C for long-term
  • Available in small research scales, e.g. 50 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.