Learn More
Medchemexpress LLC Boc-Val-Leu-Lys-AMC | 73554-84-4 | 99.0% | 615.76 | 5 MG
SDP
Catalog No. 5000397620
Encompass_Preferred
Boc-Val-Leu-Lys-AMC is a sensitive, fluorogenic, and specific substrate commonly utilized in biochemical assays. It serves as an effective substrate for plasmin, a key enzyme involved in fibrinolysis, as well as acrosin from the ascidian Halocynthia roretzi, porcine calpain isozymes I and II, and papain. This compound is valuable for researchers investigating protease activity and enzyme kinetics.
  • Sensitive fluorogenic substrate
  • Specific for plasmin and other proteases
  • Useful for studying protease activity
  • Research-grade compound
Catalog No. 50-003-97620 Supplier Medchemexpress LLC Supplier No. HYP43105MG
Boc-Val-Leu-Lys-AMC is a sensitive, fluorogenic, and specific substrate commonly utilized in biochemical assays. It serves as an effective substrate for plasmin, a key enzyme involved in fibrinolysis, as well as acrosin from the ascidian Halocynthia roretzi, porcine calpain isozymes I and II, and papain. This compound is valuable for researchers investigating protease activity and enzyme kinetics.
  • Sensitive fluorogenic substrate
  • Specific for plasmin and other proteases
  • Useful for studying protease activity
  • Research-grade compound