Learn More
Medchemexpress LLC Boc-val-pro-arg-AMC hydrochloride | 70375-24-5 | MFCD00058181 | 99.4% | 664.19 | C31H46ClN7O7 | 5 MG
SDP
Catalog No. 5000222429
Encompass_Preferred
Boc-Val-Pro-Arg-AMC hydrochloride is a sensitive fluorogenic peptide substrate used to measure trypsin-like serine protease activity. Proteolytic cleavage releases the fluorescent AMC (7-amino-4-methylcoumarin) leaving group, enabling both continuous and end-point fluorometric assays. Intended for research use only.
Catalog No. 50-002-22429 Supplier Medchemexpress LLC Supplier No. HY1377845MG
Boc-Val-Pro-Arg-AMC hydrochloride is a sensitive fluorogenic peptide substrate used to measure trypsin-like serine protease activity. Proteolytic cleavage releases the fluorescent AMC (7-amino-4-methylcoumarin) leaving group, enabling both continuous and end-point fluorometric assays. Intended for research use only.