Learn More
Carbosynth LLC. Boc-Val-Pro-Arg-MCA (5 mg vial), 1ea, MW: 627.73, Molecular formula: C31H45N7O7, CAS# 65147-04-8
SDP
Catalog No. 501686025
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Valyl-L-Prolyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Val-Pro-Arg-MCA, Boc-VPR-AMC, Boc-VPR-MCA, Purity: , Grade AA, Application: Substrate for a-Thrombin, Storage Temperature: -20C, References: T. Morita, H. Kato, S. Iwanaga, K. Takada, T. Kimura, and S. Sakakibara, J. Biochem., 82, 1495 (1977)., S. Kawabata, T. Morita, S. Iwanaga, and H. Igarashi, J. Biochem., 97, 1073 (1985).
Catalog No. 50-168-6025 Supplier Carbosynth LLC. Supplier No. MVP3093V5MG
Add to Cart
Add to Cart
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Valyl-L-Prolyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Val-Pro-Arg-MCA, Boc-VPR-AMC, Boc-VPR-MCA, Purity: , Grade AA, Application: Substrate for a-Thrombin, Storage Temperature: -20C, References: T. Morita, H. Kato, S. Iwanaga, K. Takada, T. Kimura, and S. Sakakibara, J. Biochem., 82, 1495 (1977)., S. Kawabata, T. Morita, S. Iwanaga, and H. Igarashi, J. Biochem., 97, 1073 (1985).