Learn More
Medchemexpress LLC Boc3Arg | 108787-91-3 | 97.0% | 474.55 | 25 G
SDP
Catalog No. 5000402978
Encompass_Preferred
Boc3Arg is the tert-butyl carbamate-protected arginine. It is an efficient tag to induce degradation by directly localizing the protein to the 20S proteasome.
  • Efficient tag to induce degradation
  • Directly localizes protein to the 20S proteasome
  • Target: Proteasome
  • Pathway: Metabolic Enzyme/Protease
Catalog No. 50-004-02978 Supplier Medchemexpress LLC Supplier No. 25GHY23020
Boc3Arg is the tert-butyl carbamate-protected arginine. It is an efficient tag to induce degradation by directly localizing the protein to the 20S proteasome.
  • Efficient tag to induce degradation
  • Directly localizes protein to the 20S proteasome
  • Target: Proteasome
  • Pathway: Metabolic Enzyme/Protease