Learn More
Sigma Aldrich Boca1V2Pf1-Nhpeg1-Nh2
Catalog No. 91695150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
91-695-150MG 50 mg
1 options
Catalog No. 91-695-150MG Supplier Sigma Aldrich Supplier No. 91695150MG
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Protein degrader building block BocA1V2PF1-NHPEG1-NH2 hydrochloride enables the synthesis of molecules for targeted protein degradation and SNIPER (specific and non-genetic inhibitor of apoptosis protein (IAP)-dependent protein erasers) technology. Developed in partnership with ComInnex, this conjugate contains an in silico-derived IAP-recruiting ligand, an alkyl-chain crosslinker, and a pendant amine for reactivity with an acid on a target warhead. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and protein degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, including CRBN and VHL targeted, parallel synthesis can be used to more quickly generate SNIPER and PROTAC™ degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand. Learn more about the novel IAP ligands generated through virtual screening of AVP mimetics in our Technology Spotlight. Building blocks in this series:916978 BocA1V2PF1917966 BocA1V2PF1-NHC6-NH2916706 BocA1V2PF1-NHC10-NH2916951 BocA1V2PF1-NHPEG1-NH2917214 BocA1V2PF1-NHPEG3-NH2Technology Spotlight: Degrader Building Blocks with Inhibitor of Apoptosis Protein (IAP) In Silico-Derived LigandsIn Vivo Knockdown of Pathogenic Proteins via Specific and Nongenetic Inhibitor of Apoptosis Protein (IAP)-dependent Protein Erasers (SNIPERs)SNIPERs-Hijacking IAP activity to induce protein degradationE3 Ligase Ligands for PROTACs: How They Were Found and How to Discover New OnesPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license
Specifications
Molecular Formula C30H46N4O6 -+ xHCl
Linear Formula C30H46N4O6 -+ xHCl
Quantity 50 mg
Synonym 6-((5-((6-Aminohexyl)oxy)pentyl)oxy)-N-(2-(2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-5-yl)hexanamide hydrochloride; C5 Lenalidomide conjugate; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader