Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boca1V2Pf2-Nhpeg1-Nh2

Catalog No. 91795850MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-795-850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-795-850MG Supplier Sigma Aldrich Supplier No. 91795850MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protein degrader building block BocA1V2PF2-NHPEG1-NH2 enables the synthesis of molecules for targeted protein degradation and SNIPER (specific and non-genetic inhibitor of apoptosis protein (IAP)-dependent protein erasers) technology. Developed in partnership with ComInnex, this conjugate contains an in silico-derived IAP-recruiting ligand, an alkyl-chain crosslinker, and a pendant amine for reactivity with an acid on a target warhead. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and protein degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, including CRBN and VHL targeted, parallel synthesis can be used to more quickly generate SNIPER and PROTAC™ degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand. Learn more about the novel IAP ligands generated through virtual screening of AVP mimetics in our Technology Spotlight. Building blocks in this series:916714 BocA1V2PF2917443 BocA1V2PF2-NHC6-NH2917699 BocA1V2PF2-NHC10-NH2917958 BocA1V2PF2-NHPEG1-NH2916692 BocA1V2PF2-NHPEG3-NH2Technology Spotlight: Degrader Building Blocks with Inhibitor of Apoptosis Protein (IAP) In Silico-Derived LigandsIn Vivo Knockdown of Pathogenic Proteins via Specific and Nongenetic Inhibitor of Apoptosis Protein (IAP)-dependent Protein Erasers (SNIPERs)SNIPERs-Hijacking IAP activity to induce protein degradationE3 Ligase Ligands for PROTACs: How They Were Found and How to Discover New OnesPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 154342-45-5
Molecular Formula C16H18N4O4 -+ xHCl
Linear Formula C16H18N4O4 -+ xHCl
Quantity 50 mg
Synonym 4-((3-Aminopropyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride; Crosslinker-E3 ligase ligand conjugate; Pomalidomide conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.