Learn More
Boca Scientific Inc BOCARACK800 Freezer Storage Rack Without Storage Boxes
Catalog No. NC1822334
Encompass
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include two sliding drawers and are compatible with refrigerators and -80°C freezers. The Bocarack 800 does not include tube storage boxes. It measures 24" deep x 6" wide x 10" high.
Catalog No. NC1822334 Supplier Boca Scientific Inc Supplier No. BOCARACK800
Add to Cart
Add to Cart
BOCARACK Freezer Storage Systems provide a complete solution for storing biological samples in low temperature freezers. These sturdy stainless-steel racks include two sliding drawers and are compatible with refrigerators and -80°C freezers. The Bocarack 800 does not include tube storage boxes. It measures 24" deep x 6" wide x 10" high.