Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boceprevir | 394730-60-0 | 98.5% | 519.7 g/mol | C27H45N5O5 | 100 MG
SDP

Catalog No. 5000247171
Encompass_Preferred

Boceprevir is an HCV NS3 protease inhibitor supplied as a research-grade chemical standard for biochemical and analytical applications. It is used in antiviral research and enzyme inhibition assays.

  • HCV NS3 protease inhibitor.
  • Intended for research and analytical applications.
  • Purity 98.52%.
  • CAS 394730-60-0.
  • Available as a 100 mg solid for laboratory use.

Catalog No. 50-002-47171 Supplier Medchemexpress LLC Supplier No. HY10237100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boceprevir is an HCV NS3 protease inhibitor supplied as a research-grade chemical standard for biochemical and analytical applications. It is used in antiviral research and enzyme inhibition assays.

  • HCV NS3 protease inhibitor.
  • Intended for research and analytical applications.
  • Purity 98.52%.
  • CAS 394730-60-0.
  • Available as a 100 mg solid for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.