Learn More
Medchemexpress LLC Boceprevir | 394730-60-0 | MFCD22208555 | 98.5% | 519.7 g/mol | C27H45N5O5 | 1 ML
SDP
Catalog No. 5000246430
Encompass_Preferred
Boceprevir is a peptidomimetic hepatitis C virus (HCV) NS3 protease inhibitor supplied as a pre-made 10 mM solution in DMSO for research use. The solution format simplifies dosing for biochemical assays and cell-based studies, and the compound has well-documented antiviral activity and physicochemical properties for assay development and mechanistic studies.
  • Provided as a ready-to-use 10 mM solution in DMSO.
  • Supplied in a single 1 mL vial for small-scale experiments.
  • High purity suitable for biochemical and cell-based assays.
  • Peptidomimetic protease inhibitor used in antiviral research.
  • Stable under recommended storage conditions for research reagents.
Catalog No. 50-002-46430 Supplier Medchemexpress LLC Supplier No. HY1023710MM 1ML
Boceprevir is a peptidomimetic hepatitis C virus (HCV) NS3 protease inhibitor supplied as a pre-made 10 mM solution in DMSO for research use. The solution format simplifies dosing for biochemical assays and cell-based studies, and the compound has well-documented antiviral activity and physicochemical properties for assay development and mechanistic studies.
  • Provided as a ready-to-use 10 mM solution in DMSO.
  • Supplied in a single 1 mL vial for small-scale experiments.
  • High purity suitable for biochemical and cell-based assays.
  • Peptidomimetic protease inhibitor used in antiviral research.
  • Stable under recommended storage conditions for research reagents.