Learn More
Cayman Chemical BoceprevIr-d9 500ug
SDP
Catalog No. 502992165
Encompass_Preferred
An internal standard for the quantification of boceprevir by GC- or LC-MS
Catalog No. 50-299-2165 Supplier Cayman Chemical Supplier No. 31461500 UG
An internal standard for the quantification of boceprevir by GC- or LC-MS