Learn More
Cayman Chemical BocLRRAMCtrIfluoroACTAte 10mg
SDP
Catalog No. 502933696
Encompass_Preferred
A fluorogenic substrate for trypsin-like activity of the 26S proteasome or 20S proteolytic core; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify 26S proteasome or 20S proteolytic core trypsin-like activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-3696 Supplier Cayman Chemical Supplier No. 2664210 MG
A fluorogenic substrate for trypsin-like activity of the 26S proteasome or 20S proteolytic core; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify 26S proteasome or 20S proteolytic core trypsin-like activity; ex/em = 340-360/440-460 nm, respectively