Learn More
Medchemexpress LLC BocNH-PEG5-CH2CH2Br 250mg
SDP
Catalog No. 5000205305
Encompass_Preferred
BocNH-PEG5-CH2CH2Br is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05305 Supplier Medchemexpress LLC Supplier No. 250MGHYW096092
BocNH-PEG5-CH2CH2Br is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]