Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bocnh-Peg6-Acid

Catalog No. 9026751G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-267-51G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-267-51G Supplier Sigma Aldrich Supplier No. 9026751G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This heterobifunctional, PEGylated crosslinker features a carboxylic acid at one end and Boc-protected amino group at the other, which can be deprotected with mildly acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. BocNH-PEG6-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC™ molecules) for targeted protein degradation.alphavbeta3 Integrin-Targeting Arg-Gly-Asp (RGD) Peptidomimetics Containing Oligoethylene GlycolUltrasmall particle of iron oxide-RGD peptidomimetic conjugate: synthesis and characterizationPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 1415389-64-8
Molecular Formula C112H84O8Rh2
Linear Formula C14H14F5NO2S2
Quantity 250 mg
Synonym Dirhodium tetrakis((S,R)-1-(4-phenyl(phenyl))-2,2-diphenylcyclopropanecarboxylate); Dirhodium tetrakis[(R)-1-(4-phenyl(phenyl))-2,2- diphenylcyclopropane carboxylate]; Tetrakis[(R)-1-(4-phenyl(phenyl))-2,2- diphenylcyclopropane carboxylato]dirhodium(II)
Molecular Weight (g/mol) 1763.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.