Learn More
BOD, 500 mg/L Calibration Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111004695 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
11-100-4695 500 mL
1 options
Catalog No. 11-100-4695 Supplier MilliporeSigma Supelco Supplier No. BOD500500ML
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot. Some lots may not contain all analytes.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Grade Certified Reference Material