Learn More
Hach Company BOD Bottle Brush
Supplier:  Hach Company 2985100
Encompass
Brush part 13 cm (5in ) length 5 cm (2in ) diameter Total length 33 cm (13in ) Heavily filled with black bristles on galvanized wire Radial tufted end
Catalog No. 50-002-07070