Learn More
Hach Company Bottle, BOD, 300 mL, 24/pk, Serialized: 49 - 72
Supplier: Hach Company 2898900
The standard for BOD analysis. Borosilicate glass bottle with flared lip and ground-glass stopper to prevent air entrapment and for water seal. Frosted marking area. Stopper included (60 and 300 mL sizes). Serialized bottles have silk-screened, sequential numbers and include a bar code.