Learn More
Hach Company BOD Kit HQD Series DO Meters
Catalog No. 5000207850 Shop All Hach Company Products
Encompass
BOD Kit for HQD Series DO Meters (Funnel and Magnetic Stirrer Attachment)Standard BOD Kit for HQD Series DO Meters includes funnel and magnetic stirrer attachment Requires Stir Stand (Product No 4530000 or equivalent)
Catalog No. 50-002-07850 Supplier Hach Company Supplier No. 5826600
BOD Kit for HQD Series DO Meters (Funnel and Magnetic Stirrer Attachment)Standard BOD Kit for HQD Series DO Meters includes funnel and magnetic stirrer attachment Requires Stir Stand (Product No 4530000 or equivalent)