Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ BOD Nutrient Buffer Pillows

Catalog No. 501994937 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4937 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-4937 Supplier Hach™ Supplier No. 1416066
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ready-to-use, premeasured liquid buffer concentrate in individual sealed tubes for preparation of BOD5 dilution water. Ready for immediate use.

Ready-to-use, premeasured liquid buffer concentrate in individual sealed tubes for preparation of BOD5 dilution water. Ready for immediate use.

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
For Use With (Application) Preparation of BOD5 dilution water
Includes Individual ampules
Quantity 50 Pack
Sample Size Prepares 300 mL dilution water
Size 0.5 mL ampules
Type Nutrient buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.