Learn More
Felix Storch Inc BOD Refrigerated/Heated Incubator 18 Cu.Ft. Capacity
SDP
Supplier:  Felix Storch Inc ABOD18
Encompass_Preferred
Model ABOD18 is a 15 cu.ft. refrigerated incubator. The adjustable control range allows you to manage the temperature between 4º and 60ºC (39.2 to 140ºF) and comes preset at 20ºC. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product.The current and min/max temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. The ABOD18 utilizes adjustable cycle defrost operation to avoid temperature spikes. Optimized forced air cooling with one internal fan ensures excellent temperature stability and uniformity with rapid recovery after door openings.
Catalog No. NC3901241
Add to Cart
Request a Quote