Learn More
Velp Scientifica™ BOD Sensor System 10
SDP
Catalog No. 07222012
Change view
Click to view available options
IP Rating:
IP54
Catalog No. IP Rating
07-222-012 IP54
1 options
This item is not returnable. View return policy
Catalog No. 07-222-012 Supplier Velp Scientifica™ Supplier No. SA10210137
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Innovative, mercury-free and highly reliable BOD system for BOD analysis (Biochemical Oxygen Demand) with 10 sensors and 10 positions.
  • BOD Sensor System with 10 positions
  • Determination is a simple and easy way for monitoring microorganisms' activity in water samples with highly reliable results
  • Microorganisms present or inoculated in a water sample containing biodegradable organic substance, consume oxygen for their metabolic activity and produce a corresponding volume of carbon dioxide
  • Carbon dioxide gets absorbed by a strong alkali positioned in the neck of the bottle
  • Result of this process is a decrease in gas pressure
  • BOD measures this change in gas pressure between the beginning to the end of the analysis to give a result expressed in mg/L
Specifications
Type Sensor System
IP Rating IP54
Voltage 115 V
Certifications/Compliance CE