Learn More
Hach Company BOD Standard Solution, 3000 mg/L, pk/16 - 10-mL Voluette Ampules
Catalog No. 501995140 Shop All Hach Company Products
Encompass
For accuracy checks in Biochemical Oxygen Demand (BOD) determinations.3000 mg/L Glucose plus Glutamic Acid. Pack of 16 Ampuls of 10 mL each.• Simple Accuracy Checks• Minimize waste• Reliable results
Catalog No. 50-199-5140 Supplier Hach Company Supplier No. 1486610
For accuracy checks in Biochemical Oxygen Demand (BOD) determinations.3000 mg/L Glucose plus Glutamic Acid. Pack of 16 Ampuls of 10 mL each.• Simple Accuracy Checks• Minimize waste• Reliable results