Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bode Catalyst 2 Monohydrate 98.0+%, TCI America™
SDP

Catalog No. D3983100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3983100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3983100MG Supplier TCI America Supplier No. D3983100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 903571-02-8
Molecular Formula C21H24ClN3O2
Molecular Weight (g/mol) 385.892
MDL Number MFCD10566989
InChI Key DFFDIRURHBNRTR-UITHODHBSA-M
Synonym (+)-(5aR,10bS)-5a,10b-Dihydro-2-mesityl-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium Chloride, Bode Catalyst 2
PubChem CID 54669739
SMILES CC1=CC(=C(C(=C1)C)N2C=[N+]3C4C(CC5=CC=CC=C45)OCC3=N2)C.O.[Cl-]

Specifications

Melting Point 219°C
Quantity 100 mg
Formula Weight 367.88
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.