Learn More
TARGETMOL CHEMICALS INC BODIPY 493/503 500MG
SDP
Catalog No. 502580156
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. BODIPY 493/503 (Pyrromethene 546) is a lipophilic fluorescent probe with Ex/Em of 493/503 nm. BODIPY 493/503 localizes to polar lipids and can be used to label cellular neutral lipid contents and for live and fixed cell applications. purity: 99%
Catalog No. 50-258-0156 Supplier TARGETMOL CHEMICALS INC Supplier No. T36957500MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. BODIPY 493/503 (Pyrromethene 546) is a lipophilic fluorescent probe with Ex/Em of 493/503 nm. BODIPY 493/503 localizes to polar lipids and can be used to label cellular neutral lipid contents and for live and fixed cell applications. purity: 99%