Learn More
Cayman Chemical BODIPY 493/503, Purity: ≥98%, CAS: 121207-31-6, 5 mg
SDP
Catalog No. 502932849
Encompass_Preferred
A fluorescent probe for lipid droplets; can be used to label cellular neutral lipid contents, particularly those localized to lipid droplets, in live and fixed cells; compatible with epifluorescent, confocal, and two-photon microscopy, as well as flow cytometry; ex/em = 493/503 nm
Catalog No. 50-293-2849 Supplier Cayman Chemical Supplier No. 258925 MG
A fluorescent probe for lipid droplets; can be used to label cellular neutral lipid contents, particularly those localized to lipid droplets, in live and fixed cells; compatible with epifluorescent, confocal, and two-photon microscopy, as well as flow cytometry; ex/em = 493/503 nm