Learn More
Cayman Chemical BODIPY 505/515, Purity: ≥98%, CAS: 21658-70-8, 250 mg
SDP
Catalog No. 502934964
Encompass_Preferred
A fluorescent probe for lipid droplets; has been used for electron, epifluorescent, and confocal microscopy, as well as flow cytometry applications; has been used for live and fixed cell applications; ex/em = 505/515
Catalog No. 50-293-4964 Supplier Cayman Chemical Supplier No. 25893250 MG
A fluorescent probe for lipid droplets; has been used for electron, epifluorescent, and confocal microscopy, as well as flow cytometry applications; has been used for live and fixed cell applications; ex/em = 505/515