Learn More
Cayman Chemical BODIPY 508CholSTRyl Ol
SDP
Catalog No. 502919271
Encompass_Preferred
A fluorescently tagged cholesterol ester; ex/em = 480/508 nm, respectively; has been used to monitor cholesterol trafficking in HeLa cells
Catalog No. 50-291-9271 Supplier Cayman Chemical Supplier No. 39685500 UG
A fluorescently tagged cholesterol ester; ex/em = 480/508 nm, respectively; has been used to monitor cholesterol trafficking in HeLa cells