Learn More
Cayman Chemical BODIPY 508CholSTRyl Ol
Supplier: Cayman Chemical 39685250 UG
A fluorescently tagged cholesterol ester; ex/em = 480/508 nm, respectively; has been used to monitor cholesterol trafficking in HeLa cells