Learn More
Medchemexpress LLC BODIPY 558/568 C12 | 158757-84-7 | 98.4% | 472.40 | 1 ML
SDP
Catalog No. 5000347782
Encompass_Preferred
BODIPY 558/568 C12 is a BODIPY dye, a small molecule dye known for strong ultraviolet absorption, a relatively sharp fluorescence peak, and high quantum yield. It is relatively insensitive to environmental polarity and pH, and stable under various physiological conditions. Due to its structural asymmetry, it yields diverse structural products. BODIPY lipid droplet dyes can penetrate cell membranes, localizing neutral lipids within cells to specifically stain lipid droplets.
  • Strong ultraviolet absorption
  • Relatively sharp fluorescence peak
  • High quantum yield
  • Relatively insensitive to environmental polarity and pH
  • Stable under various physiological conditions
  • Suitable for labeling live and fixed cells
  • Maximum excitation/emission wavelength: 558/568 nm
Catalog No. 50-003-47782 Supplier Medchemexpress LLC Supplier No. HY13822610MM/1ML
BODIPY 558/568 C12 is a BODIPY dye, a small molecule dye known for strong ultraviolet absorption, a relatively sharp fluorescence peak, and high quantum yield. It is relatively insensitive to environmental polarity and pH, and stable under various physiological conditions. Due to its structural asymmetry, it yields diverse structural products. BODIPY lipid droplet dyes can penetrate cell membranes, localizing neutral lipids within cells to specifically stain lipid droplets.
  • Strong ultraviolet absorption
  • Relatively sharp fluorescence peak
  • High quantum yield
  • Relatively insensitive to environmental polarity and pH
  • Stable under various physiological conditions
  • Suitable for labeling live and fixed cells
  • Maximum excitation/emission wavelength: 558/568 nm